Gold ≥99.99%, disk, as rolled, Diameter 6 mm

Supplier: GOODFELLOW CORPORATION

1000030673 1000013900 1000030704 1000032087 1000030569 1000031587
77680-855EA 250.98 USD
77680-855 77680-703 77680-867 77681-180 77680-826 77681-050
Gold ≥99.99%, disk, as rolled, Diameter 6 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR