Gold ≥99.99%, disk, as rolled, Diameter 30 mm
Supplier: GOODFELLOW CORPORATION
1000030664
1000031915
1000030696
1000030561
1000031571
1000032078
77680-850EA
617.97
USD
77680-850
77681-132
77680-863
77680-820
77681-044
77681-174
Gold ≥99.99%, disk, as rolled, Diameter 30 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...