Gold ≥99.9%, disk, Diameter 6 mm

Supplier: GOODFELLOW CORPORATION

1000030324 1000030426 1000030380 1000030531 1000030943 1000030982 1000031029 1000031338 1000031186 1000031753 1000030474 1000031837 1000030829 1000031649 1000031795
77680-745EA 139.73 USD
77680-745 77680-786 77680-771 77680-812 77680-920 77680-928 77680-937 77681-011 77680-976 77681-083 77680-799 77681-109 77680-898 77681-064 77681-094
Gold ≥99.9%, disk, Diameter 6 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR