Gold ≥99.9%, disk, Diameter 12.5 mm

Supplier: GOODFELLOW CORPORATION

1000030403 1000030360 1000030967 1000030513 1000031007 1000030924 1000031171 1000031773 1000030304 1000030793 1000031614 1000031732 1000030456 1000031323 1000031818
77680-774EA 213.26 USD
77680-774 77680-759 77680-923 77680-801 77680-931 77680-915 77680-971 77681-085 77680-733 77680-886 77681-053 77681-073 77680-789 77681-006 77681-097
Gold ≥99.9%, disk, Diameter 12.5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR