Gold ≥99.9%, disk, Diameter 25 mm

Supplier: GOODFELLOW CORPORATION

1000030412 1000030370 1000030521 1000030314 1000030464 1000031629 1000031826 1000030808
77680-779EA 486.99 USD
77680-779 77680-764 77680-806 77680-738 77680-794 77681-058 77681-102 77680-891
Gold ≥99.9%, disk, Diameter 25 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR