Gold ≥99.999%, disk, as rolled, Diameter 12.5 mm

Supplier: GOODFELLOW CORPORATION

1000030335 1000031113 1000031376 1000031865 1000031215
77680-748EA 2439.11 USD
77680-748 77680-956 77681-014 77681-112 77680-979
Gold ≥99.999%, disk, as rolled, Diameter 12.5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR