Gold ≥99.9%, disk, Diameter 4 mm
Supplier: GOODFELLOW CORPORATION
1000030319
1000030420
1000030373
1000030976
1000030526
1000031023
1000030937
1000031180
1000031746
1000030470
1000031831
1000030817
1000031332
1000031637
1000031786
77680-741EA
128.33
USD
77680-741
77680-782
77680-767
77680-926
77680-809
77680-935
77680-918
77680-974
77681-080
77680-797
77681-105
77680-894
77681-009
77681-060
77681-091
Gold ≥99.9%, disk, Diameter 4 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...