Gold ≥99.9%, disk, Diameter 30 mm

Supplier: GOODFELLOW CORPORATION

1000030416 1000030371 1000014342 1000030522 1000031781 1000030315 1000030466 1000031741 1000031828 1000030811
77680-780EA 486.99 USD
77680-780 77680-765 77680-704 77680-807 77681-089 77680-739 77680-795 77681-078 77681-103 77680-892
Gold ≥99.9%, disk, Diameter 30 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR