Gold ≥99.95%, disk, as rolled, Diameter 10 mm

Supplier: GOODFELLOW CORPORATION

1000030860 1000031935 1000007427 1000024021 1000030722 1000031050 1000031986 1000032096 1000030615 1000031698
77680-900EA 398.7 USD
77680-900 77681-137 77680-699 77680-724 77680-869 77680-939 77681-153 77681-182 77680-828 77681-066
Gold ≥99.95%, disk, as rolled, Diameter 10 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR