Gold ≥99.95%, disk, as rolled, Diameter 25 mm

Supplier: GOODFELLOW CORPORATION

1000030740 1000030878 1000002782 1000031068 1000031994 1000031280 1000032104 1000030624 1000031704 1000031945
77680-875EA 895.78 USD
77680-875 77680-906 77680-692 77680-945 77681-158 77680-998 77681-187 77680-833 77681-068 77681-143
Gold ≥99.95%, disk, as rolled, Diameter 25 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR