NanoXact Mesoporous Silica Nanospheres 5 /mg/ml in Ethanol, nanoComposix
Supplier: Avantor
SHSN100-100M SHSN100-30M SHSN100-5M
103820-568EA 1430 USD
103820-568 77680-568 77680-570 77680-572
NanoXact Mesoporous Silica Nanospheres 5 /mg/ml in Ethanol, nanoComposix
Beads Microcarriers
High quality mesoporous silica nanoparticles, provided with full COA
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Silica surface offers robust and versatile surface for a wide variety of applications
Spherical mesoporous silica nanoparticles with a silanol surface. Like sponges, mesoporous silica nanoparticles (MSNs) can adsorb exceptionally large quantities of molecules in their interior spaces (pores) thanks to their high surface area. High quality mesoporous silica particles with uniform pore sizes and consistent pore structure can act as “advanced nanosponges” by only adsorbing molecules within a specific size range into their cavities. Uses for mesoporous silica range from catalysis and environmental chemical removal to biomedicine where they are widely used as drug delivery system (DDS). Their high surface area, easy surface modification, and biocompatibility make them the material of choice for targeted delivery of active pharmaceutical ingredients for cancer therapy and other therapies and treatments.
Each batch is extensively characterized to ensure it meets the highest quality standards so you can achieve reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures
Caution: Do not freeze. Store at 25 °C, bath sonicate if needed. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.