NanoXact Gold Nanorods, 1 or 50 OD, Citrate, PEG, or Carboxyl Surface, 660, 800 or 980 nm Peak Resonance, nanoComposix
Supplier: Avantor
GRCN980-1L GRCN980-25M GRCN980-500M GRGH660-15M GRGH660-1M GRGH660-500U GRGH660-5M GRGH800-15M GRGH800-1M GRGH800-500U GRCH660-15M GRCH660-1M GRCH660-500U GRCH800-15M GRCH800-1M GRCH800-500U GRCH800-5M GRCH980-15M GRCH980-1M GRCH980-500U GRCH980-5M GRCN660-100M GRCN660-1L GRCN660-25M GRCN660-500M GRCN800-100M GRCN800-1L GRCN800-25M GRCN800-500M GRCN980-100M GRGH800-5M GRXH660-15M GRXH660-1M GRXH660-500U GRXH660-5M GRXH800-15M GRXH800-500U GRXH800-5M
103369-588EA 215 USD
103369-588 103369-964 103369-968 103369-970 103820-164 103820-166 103820-168 103820-170 103820-172 103820-174 103820-176 103820-178 103820-188 103820-190 103820-200 103820-202 103820-204 103820-206 103820-208 103820-210 103820-212 103820-214 103820-216 103820-218 103820-220 103820-222 103820-224 103820-226 103820-228 103820-230 77680-282 77680-284 77680-286 77680-288 77680-290 77680-292 77680-294 77680-296 77680-298 77680-300 77680-240 77680-242 77680-244 77680-246 77680-248 77680-250 77680-252 77680-254 77680-256 77680-258 77680-260 77680-262 77680-264 77680-266 77680-268 77680-270 77680-272 77680-274 77680-276 77680-278 77680-280 77680-302 77680-304 77680-306 77680-308 77680-310 77680-312 77680-314 77680-316 77680-318
NanoXact Gold Nanorods, 1 or 50 OD, Citrate, PEG, or Carboxyl Surface, 660, 800 or 980 nm Peak Resonance, nanoComposix
Beads Microcarriers
Gold nanorods offered at 1 OD or 50 OD (high) concentration with peak resonance options of 660 nm, 800 nm, and 980 nm.
  • For use in diagnostics, photothermal therapies, imaging, drug delivery, and more.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • CTAB-free
  • Citrate surface is stable and displaceable
  • PEG surface offers biocompatibility
  • Carboxyl surface is ideal for conjugation
The Citrate surface is stable and displaceable for easy functionalization with silica, DNA, and antibodies. The PEG surface offers high stability, compatibility with some alcohols, and biocompatibility. The Carboxyl surface can be used for conjugation to proteins, antibodies, and other biomolecules via EDC/NHS chemistry.
All gold nanorods from nanoComposix are CTAB-free and provided in water. Many gold nanorods available commercially are coated with cetyl trimethylammonium bromide (CTAB), a common surfactant used during nanorod synthesis. CTAB coated nanorods are toxic in biological systems and are readily destabilized when transferred to different solvents. NanoComposix’s gold nanorods are CTAB-free and highly purified to remove residual reactants following synthesis. Gold nanorods have used in optical filters, rapid diagnostics like Lateral Flow Immunoassays, sensing and biosensing devices, photothermal therapies, and many other applications.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, and UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Ordering information: Custom formulations and ISO13485/cGMP compliant materials can be provided on request.