BioPure Gold Nanospheres, 1 mg/ml, Citrate Surface, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AUCB10-5M AUCB10-10M AUCB20-1M AUCB20-5M AUCB20-10M AUCB30-1M AUCB30-5M AUCB30-10M AUCB40-1M AUCB40-5M AUCB40-10M AUCB50-1M AUCB50-5M AUCB50-10M AUCB60-1M AUCB60-5M AUCB60-10M AUCB80-1M AUCB80-5M AUCB80-10M AUCB100-1M AUCB100-5M AUCB100-10M AUCB5-1M AUCB5-5M AUCB5-10M AUCB10-1M AUCB20-10M AUCB20-1M AUCB20-5M AUCB30-10M AUCB30-1M AUCB30-5M AUCB40-10M AUCB40-1M AUCB60-10M AUCB60-1M AUCB60-5M AUCB80-10M AUCB80-1M AUCB80-5M AUCB100-10M AUCB100-1M AUCB100-5M AUCB10-10M AUCB10-1M AUCB10-5M AUCB40-5M AUCB50-10M AUCB50-1M AUCB50-5M AUCB5-10M AUCB5-1M AUCB5-5M
103369-238EA 1220 USD
103369-238 103369-240 103369-248 103369-250 103369-252 103369-254 103369-256 103369-258 103369-260 103369-262 103369-264 103369-266 103369-268 103369-270 103369-272 103369-274 103369-276 103369-734 103369-736 103369-738 103369-740 103369-742 103369-744 103369-902 103369-904 103369-906 103369-914 77679-902 77679-904 77679-908 77679-910 77679-912 77679-916 77679-918 77679-920 77679-942 77679-944 77679-948 77679-950 77679-952 77679-956 77679-886 77679-888 77679-892 77679-894 77679-896 77679-900 77679-924 77679-926 77679-928 77679-932 77679-934 77679-936 77679-940
BioPure Gold Nanospheres, 1 mg/ml, Citrate Surface, nanoComposix
Beads Microcarriers
Purified, concentrated, high quality gold nanoparticles. Sterile and endotoxin-free. Bare citrate surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/mL, 5.08 mM, 1000 ppm, ~16 to 30 OD)
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra-pure (Milli-Q) DI water
  • BioPure materials ideal for in-vitro and in-vivo experiments, nanotoxicology
  • "Bare" citrate surface is stable and displaceable
Spherical gold nanoparticles, 1 mg/mL (high concentration)
Each batch is extensively characterized to meet the highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Accessories information: Accessories not included.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included. Due to the temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: Do not freeze. Store at 2 to 8°C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.