BioPure Silver Nanospheres 1 mg/ml PVP (polyvinylpyrrolidone for high stability), nanoComposix
Supplier: NANOCOMPOSIX, INC.
AGPB100-10M AGPB100-1M AGPB100-30M AGPB100-5M AGPB50-10M AGPB50-1M AGPB50-30M AGPB50-5M AGPB50-10M AGPB50-1M AGPB50-30M AGPB50-5M
103819-668EA 1865 USD
103819-668 103819-670 103819-672 103819-674 103819-700 103819-702 103819-704 103819-706 77679-806 77679-808 77679-810 77679-812
BioPure Silver Nanospheres 1 mg/ml PVP (polyvinylpyrrolidone for high stability), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality silver nps; sterile and endotoxin-free. PVP for stable surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 9.27 mm, 1000 ppm, ~40 to 135 OD [depending on size])
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • PVP surface provides strong steric stability
Spherical silver nanoparticles, 1 mg/ml (high concentration). PVP polymer surface provides strong steric stability (40 kD polyvinylpyrrolidone). BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <15% for 30 to 200 nm sizes, <20% for 20 nm, and <25% for 5 and 10 nm sizes).
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: Do not freeze. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.