Harvard Apparatus Pump 11 Elite Syringe Pumps
Supplier: INSTECH LABORATORIES, INC
HA1100
77679-318EA 3692.05 USD
77679-318
Harvard Apparatus Pump 11 Elite Syringe Pumps
Pumps Syringe Pumps
The Pump 11 Elite series expands the capabilities of the original workhorse Pump 11 series with a high resolution touchscreen and intuitive icon interface. The interface allows users to create, save, and run simple to complex methods without a PC.
  • Easy to use with an LCD color touchscreen and icon interface
  • Outstanding flow performance
  • Easily create, save, and run simple to complex methods without a PC
  • Alphanumeric keypad for easy method naming and recall
  • Adjustable linear force up to 35 lbs.
  • Upgrade new versions of software remotely
These syringe pumps have a mechanism that includes a tight gripping, more secure syringe clamp for syringes ranging from 0.5 µl to 60 ml (single syringe) and 0.5 µl to 10 ml (dual syringe). The Pump 11 Elite series offers enhanced flow performance with high accuracy and smooth flow from 0.58 picoliters/min. up to 88.28 ml/min., depending on the model and syringe size.