VWR® Standard Glass Door Laboratory Refrigerator 20 cu. ft.
Supplier: VWR International
77677-320
77677-320EA 4382.56 USD
77677-320
VWR® Standard Glass Door Laboratory Refrigerator 20 cu. ft.
Refrigerators
The VWR® Laboratory Refrigerators are versatile cold storage units which deliver superior temperature control to laboratory environments.
  • Superior temperature uniformity
  • Microprocessor temperature controllers
  • Audible and visual high and low temperature alarms
  • Forced draft air circulation
  • Natural hydrocarbon refrigerants
These VWR® Laboratory Refrigerators are specifically designed for scientific and clinical use. These refrigerators feature a digital microprocessor temperature controller, ensuring precise temperature management for critical samples and supplies.
In addition, a forced air directional refrigeration system provides excellent temperature uniformity and rapid recovery after door openings. Upgrade your laboratory with these versatile units.
Certifications: ETL/cETL Listed.