Bench Top Biohazard Waste Container
Supplier: CP Lab Safety
SWC-002BIO
77677-218EA 134.84 USD
77677-218
Bench Top Biohazard Waste Container
Bins Waste Bins
Bench top biohazard waste container with lid is a convenient way to discard of contaminated biohazardous wastes right where you work. This product is the same as solid waste container but comes with the universal Biohazard label affixed to the container to clearly indicate its contents.
  • This waste container has a small footprint and can easily sit in the corner of a fume hood or bench
  • Made of chemically resistant HDPE and features a detachable base
Features a neon red-orange biohazard label to clearly indicate the nature of its contents. Featuring a detachable base and lid with gasket, our Biohazard waste container is an ideal containment system for contaminated solid wastes, preventing possible cross contamination of products.
The latching lid significantly reduces the emission of biohazardous waste back into the workspace. Ideal if you routinely handle blood borne pathogens, contaminated gloves, wipes, pipets, and other biohazardous disposables.
Comes with a single sample liner bag of 45.72 cm (18") length, 35.56 cm (14") width, 0.05 mm (thick).
Footprint: 13×13" (with base), 7×7" (without base).