Drum Lock Safety Clamp Starter Kits
Supplier: CP Lab Safety
CFS-3000_1117-YELLOW CFS-3000_1118-BLACK CFS-3000_1118-BLUE CFS-3000_1118-GREEN CFS-3000_1118-ORANGE CFS-3000_1118-RED CFS-3000_1118-YELLOW CFS-3000_1119-BLACK CFS-3000_1119-BLUE CFS-3000_1119-GREEN CFS-3000_1119-ORANGE CFS-3000_1119-RED CFS-3000_1119-YELLOW CFS-3000_1120-BLACK CFS-3000_1120-BLUE CFS-3000_1120-GREEN CFS-3000_1120-ORANGE CFS-3000_1120-RED CFS-3000_1120-YELLOW CFS-3000_1117-BLACK CFS-3000_1117-BLUE CFS-3000_1117-GREEN CFS-3000_1117-ORANGE CFS-3000_1117-RED
77676-634EA 61.39 USD
77676-634 77676-636 77676-638 77676-640 77676-642 77676-644 77676-646 77676-648 77676-650 77676-652 77676-654 77676-656 77676-658 77676-660 77676-662 77676-664 77676-666 77676-668 77676-670 77676-624 77676-626 77676-628 77676-630 77676-632
Drum Lock Safety Clamp Starter Kits
Clamps General Purpose Clamps
Drum lock safety clamp starter kit provides everything you need to safely feed chemicals in and out of drums while minimizing fumes and complying with FDA regulations.
  • Fit standard plastic UN chemical drums between 15-gals. and 55-gals. that feature a 2" opening with a flange
  • Clamps can be applied or removed at any time during fluid feed operation
  • Use in conjunction with the chemical drum seal
  • Includes one EPDM tapered plug and one silicone plug for extreme chemicals such as chloroform, bromine, and carbon tetrachloride
A safe, fast, effective way to increase the safety of your chemical feed system. Outer drum-lock clamp housing is made of polypropylene and is available in many colors to reduce cross-contamination of chemicals, by color-coding safety devices to chemical drums. Ideal for anyone who uses chemical feeds and Clean-in-place (CIP) methods.
Benefits:
Easy to use and cost-effective solution to Chemical feed problems
Reduces risk of direct chemical exposure due to 'splashing'
Improves 'Indoor Air Quality' (IAQ) - helpful for OSHA inspections
Reduces contamination of the chemical product by helping to eliminate any dirt, debris, moisture, or environmental hazards from entering the drum
Aids to reduce cross-contamination by using color-coded safety clamps on chemical drums
Creates a 'greener' work environment, which aids employee's health
Solid filter and drum seal tags enable proper Lock out, Tag out procedure for safe storage of chemicals
Applications:
Beverage processing, institutional, hospitality, pharmaceuticals, chemical processing, power generation utilities, refining, mining, ethanol, food manufacturing, semi-conductor, pulp and paper, health care, laundry, design and build engineering services, contract manufacturing.
Delivery information: 1× Drum feed safety clamp, 1× Replacement filter, 1× Spare EPDM plug fitting CFS-1013, 1× Spare Silicone plug fitting CFS-1012, 10× Drum seal tags # CFS-1014 (lock-out tag-out serialized ID safety tags).