Lockout/Tagout LOTO Drum Lock Safety Clamps with Solid Filters
Supplier: CP Lab Safety
CFS-1120-BLACK CFS-1120-BLUE CFS-1120-GREEN CFS-1120-ORANGE CFS-1120-RED CFS-1120-YELLOW
77676-612EA 48.47 USD
77676-612 77676-614 77676-616 77676-618 77676-620 77676-622
Lockout/Tagout LOTO Drum Lock Safety Clamps with Solid Filters
Security Locks
Drum Lock Safety Clamp with Solid Venting Chemical Filter, Only for use with plastic drums. Do not use with metal drums. Use in conjunction with the Drum Seal Tags (Part #CFS-1014_10pk) for Lock-out, Tag-out (LOTO) of a chemical drum, while off-line (not included.) A safe, fast, effective way to increase the safety of your chemical feed system. Outer drum-lock clamp housing is made of polypropylene and is available in many colors to reduce cross-contamination of chemicals, by color-coding safety devices to chemical drums. This clamp is used when you want to store a chemical drum that has been opened and the opening needs to be closed. You cannot feed tubing through this drum clamp.
  • Easy to use and cost-effective solution to Chemical Feed Problems
  • Reduces risk of direct chemical exposure due to 'splashing'
  • Improves 'Indoor Air Quality' (IAQ) - helpful for OSHA inspections
  • Reduces contamination of the chemical product by helping to eliminate any dirt, debris, moisture, or environmental hazards from entering the drum
  • Aids to reduce cross-contamination by using color-coded Safety Clamps on chemical drums
  • Creates a 'greener' work environment, which aids employee's health
  • Solid Filter and Drum Seal Tags enable proper Lock out, Tag out procedure for safe storage of chemicals
  • Closure size: 2"
Industries to use: Beverage processing dairy, brewery/distillation, water/soda/etc., institutional, general water treatment, pools and spas, car washes, hospitality, pharmaceuticals, chemical processing, power generation/utilities, refining, mining, ethanol, food manufacturing, semi-conductor, pulp and paper, health care, laundry , design and build engineering services and contract manufacturing.