Drum Lock Safety Clamps, Tear-Out Center
Supplier: CP Lab Safety
CFS-1119-BLACK CFS-1119-BLUE CFS-1119-GREEN CFS-1119-ORANGE CFS-1119-RED CFS-1119-YELLOW
77676-600EA 48.47 USD
77676-600 77676-602 77676-604 77676-606 77676-608 77676-610
Drum Lock Safety Clamps, Tear-Out Center
Security Locks
Drum lock safety clamp, enhanced All-In-One flexible hole size. A safe, fast, effective way to increase the safety of your chemical feed system.
  • Easy to use and cost-effective solution to chemical feed problems
  • Reduces risk of direct chemical exposure due to 'splashing'
  • Improves 'Indoor Air Quality' (IAQ) - helpful for OSHA inspections
  • Reduces contamination of the chemical product by helping to eliminate any dirt, debris, moisture, or environmental hazards from entering the drum
  • Aids to reduce cross-contamination by using color-coded Safety Clamps on chemical drums.
  • Creates a 'greener' work environment, which aids employee's health
  • Solid Filter and Drum Seal Tags enable proper Lock out, Tag out procedure for safe storage of chemicals
Outer drum-lock clamp housing is made of polypropylene and is available in many colors to reduce cross-contamination of chemicals, by color-coding safety devices to chemical drums. Ideal for anyone who uses chemical feeds and Clean-in-place (CIP) methods. Only for use with plastic drums. Do not use with metal drums.
As a general rule, these drum locks fit standard plastic UN chemical drums between 15 gallon and 55 gallon that feature a 2" opening with a flange. The precise size range that it will fit is 85 - 92 mm. Filter fits all sizes of feed hoses, tubes, etc. and is the most versatile drum lock in terms of tubing size (fits up to 1.2" tubing with all inner rings removed.) Custom-fit filter allows the end user to tear away various center sections until the opening fits their feed tube apparatus.
Memory properties allow the enhanced filter to close around the feed tube or probe and retain perfect shape for the next drum change-out. Recommended for all water treatment applications, including where a high-strength acid or base are involved. Recommended for cleaning and sanitation applications, including feed systems where multiple tubes and/or probes are being used in the same bung opening. Clamp can be applied or removed at any time during chemical feed operation. Can be used in conjunction with a chemical drum seal tag, for compliance with the FDA FSMA (Food Safety Modernization Act). It allows you to seal a chemical drum whilefeeding product.
Delivery information: Includes 1 black EPDM tapered plug and cross-linked, closed-cell, copolymer filter that provides optimum chemical resistance.