Neutral red 3.33 g/l, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

115-341-061
77676-552EA 35.92 USD
77676-552
Neutral red 3.33 g/l, sterile filtered
Neutral red

Neutral Red Solution is also known as Basic Red 5, C.I. 50040, toluylene red, or 3-amino-7-dimethyl amino-2-methylphenazine HCl.
This formulation of neutral red contains 3.33 g/L of the cationic neutral red dye.


Applications:

Histology: Neutral red is commonly used in histological staining applications. (Cell Staining Protocol and Gram Staining Protocol)

Virology: The neutral red solution is also used in the plaque assay for the quantification of infectious particles by quantifying the plaques formed in cell culture following infection with serial dilutions of an infectious sample. (Plaque Assay Protocol)

Cytology: The neutral red solution application extends to the assessment of cell proliferation and cytotoxicity in the neutral red uptake assay used to quantify the number of viable cells in culture. (Neutral Red Uptake (NRU) Assay)


Caution: Max Shelf Life - 18 months from date of manufacture

Formula: C₁₅H₁₇ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD00012651
CAS Number: 553-24-2

Order Now

Specification Test Results

Form Red liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR