AccuTec Pro® Single Edge Blades - Round Corner, AccuTec Blades
Supplier: Accutec Blades
APBL-7041-0000
77676-540CS 637.47 USD
77676-540
AccuTec Pro® Single Edge Blades - Round Corner, AccuTec Blades
Razors and Blades
AccuTecPro Single Edge blades feature precision ground edges for sharpness and durability. Blades are unwrapped for high volume use.
  • Round corner design for safety
  • 2-facet edge for increased durability
  • Heavy duty .012" carbon steel
  • Bulk, unwrapped blades are ideal for high volume use
  • Each box contains 225 blades
  • Made in USA
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.