Ergonomic Stereo Microscope for Inspection, M80, Leica Microsystems
Supplier: Leica Microsystems
8122687
77676-438EA 13520.78 USD
77676-438
Ergonomic Stereo Microscope for Inspection, M80, Leica Microsystems
Microscopes
The Leica M80 stereo microscope: Ergonomic, precise, and designed for professionals. With Leica optics and user-friendly features, it ensures efficiency and comfort during prolonged usage. Elevate your inspection experience with the Leica M80, where precision meets ergonomic design.
  • 8:1 zoom range, with a continuous zoom from 7.5× to 60× including engageable click stops
  • Leica optics that deliver outstanding resolution paired with great depth of field, so that you see all relevant structures at a glance
The Leica M80 stereo microscope sets the standard for precision inspection with its ergonomic design. Crafted for professionals, this microscope offers Leica optics and a comfortable ergonomic setup, ensuring prolonged usage without compromising efficiency. The Leica M80 seamlessly integrates clarity and user-friendly features, making it the smart choice for professionals seeking an ergonomic solution for detailed inspection tasks. Elevate your inspection experience with the Leica M80, where precision meets ergonomic design for optimal performance in the field of microscopy.
Features and specifications include:
Leica M80 optics carrier with zoom magnification changer 8:1
Magnification range: 7.5× to 60× (1× objective, 10× eyepieces)
Optics parfocal
Eight click stop zoom settings
Modular system
Suitable for achromatic, plan or planapo objectives
Objective thread M60
ESD
Delivery information: Includes Leica M80 optics carrier; eyepiece 10×/23B, adjustable, 3d gen x²; ergo tube 10 to 50°, binocular; objective achromat 1.0×, WD = 90 mm; power cable, 2 m, USA; focus drive 500 mm M-series routine; microscope carrier; Inc. light base, large w. antiShock feet; intermediate adapter for TL and IL base; Leica LED3000 RL, 58 mm; polarisation set for LED3000 RL; power supply for LED3000/LED5000; dust cover antistatic, small.