Ergonomic Stereo Microscope for Inspection with Removable Integrated Camera, M80, Leica Microsystems
Supplier: Leica Microsystems
8122686
77676-436EA 17345.05 USD
77676-436
Ergonomic Stereo Microscope for Inspection with Removable Integrated Camera, M80, Leica Microsystems
Microscopes
The Leica M80 stereo microscope offers users exceptional clarity and ease of use. Its ergonomic design ensures comfort during prolonged use. With a removable integrated camera, it allows for documentation and analysis. Elevate your inspection experience with the Leica M80 for optimal performance.
  • 8:1 zoom range, with a continuous zoom from 7.5× to 60× including engageable click stops
  • Leica optics that deliver outstanding resolution paired with great depth of field, so that you see all relevant structures at a glance
  • The removable integrated camera adds a layer of flexibility, allowing users to document and analyse their inspections seamlessly
The Leica M80 stereo microscope is a versatile solution, excelling in inspection tasks with its ergonomic design and removable integrated camera. This microscope offers unparalleled clarity and ease of use, enhancing the efficiency of inspection processes. Its ergonomic features ensure comfortable use during prolonged periods of work. The removable integrated camera adds a layer of flexibility, allowing users to document and analyze their inspections seamlessly. Elevate your inspection experience with the Leica M80, a reliable stereo microscope that combines precision, ergonomics, and advanced imaging capabilities for optimal performance.
Features and specifications include:
Leica M80 optics carrier with zoom magnification changer 8:1
Magnification range: 7.5× to 60× (1× objective, 10x eyepieces)
Optics parfocal
Eight click stop zoom settings
Modular system
Suitable for achromatic, plan or planapo objectives
Objective thread M60
ESD
Delivery information: Includes Leica M80 optics carrier; eyepiece 10×/23B, adjustable, 3d gen x²; ergo tube 10 to 50°, binocular; objective achromat 1.0×, WD = 90 mm; power cable, 2 m, USA; focus drive 500 mm M-series routine; microscope carrier; Inc. light base, large w. antiShock feet; intermediate adapter for TL and IL base; Leica LED3000 RL, 58 mm; polarisation set for LED3000 RL; power supply for LED3000/LED5000; dust cover antistatic, small; flexacam i5 (stereo); samsung 4k HDMI monitor.