The Leica Ivesta 3 stereo microscope: a precise instrument for meticulous inspection and rework. With Leica optics, ergonomic design, and advanced features, it's an essential tool for professionals. Elevate workmanship, accuracy, and efficiency with Leica Ivesta 3.
- Change from sample overview to detail easily with the microscope's large 9:1 zoom range
- 122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
- Save time refocusing as you can see every important detail at a glance
- FusionOptics allows you to perceive your samples in 3D with up to 3× larger depth of field
- Reveal fine details with a magnification of up to 55x and apochromatically corrected optics
The Leica Ivesta 3 stereo microscope is a precision instrument engineered for meticulous inspection and rework tasks. Designed for professionals, this microscope offers Leica optics, ensuring unparalleled clarity in detailed work. Its ergonomic design and advanced features make it an indispensable tool for professionals engaged in inspection and rework applications. Elevate your workmanship with the Leica Ivesta 3, a reliable stereo microscope crafted to enhance accuracy and efficiency in tasks requiring meticulous attention and precision.
Features and specifications include:
9:1 zoom with magnification 6.1× to 55× (10× eyepieces, no supplementary lens) - provides field of view up to 37.6 mm for fast, accurate inspection
12 mm depth of field with fusion optics. Greenough optical design - provides very flat (plan) field of view and large depth of field
Complete apochromatic corrected microscopy system - 35° angle eye tubes - for comfort and convenience
122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
Resolution up to 250 lp/mm (without supplementary lens, 10× eyepieces), maximum resolution up to 500 lp/mm (2× supplementary lens, 10x eyepieces)
Antimicrobial PC/ABS housing with AgTreat™
Clickstops at 10× 20× 30× 40× and 50× – inspection at specific magnification can be easily repeated
Large range of supplementary lenses as well as eyepieces
Leica LED2500, LED stand with incident and transmitted light, For S-M- and DMS-Series 6100K colour temperature four-point ring light for high brightness 3-point arc light for oblique illumination.
Transmitted light with centered LED for contrast enhancing integrated power supply: 100 to 240 V
Dimension of Base : 331×257 mm
Height of base: 35 mm
Adjustable focus torque
Delivery information: Includes Ivesta 3; eyepiece 10×/23B, adj., eyeglasses x²; power cable, 2 m, USA; Leica LED2500; dust cover antistatic, small.