Digital Microscopes for Inspection, Emspira 3
Supplier: Leica Microsystems
8122665
77676-400EA 6270.95 USD
77676-400
Digital Microscopes for Inspection, Emspira 3
Microscopes
The Leica Ivesta 3 stereo microscope is a tool designed for sorting. It offers clear optics, user-friendly features, and an integrated camera for documentation. Streamline your workflow with this efficient and reliable microscope for daily lab tasks.
  • Change from sample overview to detail easily with the microscope's large 9:1 zoom range
  • 122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
  • Save time refocusing as you can see every important detail at a glance
  • FusionOptics allows you to perceive your samples in 3D with up to 3x larger depth of field
  • Reveal fine details with a magnification of up to 55× and apochromatically corrected optics
  • Enable fast and easy discussions of results to get other opinions: Fast display of live images from the 12-megapixel camera on a monitor
  • Visualize sample movements on the monitor clearly and smoothly thanks to the high frame rate of up to 60 fps
The Leica Ivesta 3 stereo microscope is the perfect solution for routine sorting applications, combining precision optics with user-friendly features. This microscope not only offers exceptional clarity and ease of use but also includes an integrated camera for documentation purposes. With advanced capabilities and convenient documentation features, the Leica Ivesta 3 streamlines routine cell sorting processes, providing researchers and professionals with a reliable and efficient tool for daily laboratory tasks. Elevate your workflow and capture important moments seamlessly with the performance and convenience offered by the Leica Ivesta 3 stereo microscope and its integrated camera for documentation.
Features and specifications include:
9:1 zoom with magnification 6.1 to 55× (10× eyepieces, no supplementary lens) - provides field of view up to 37.6 mm for fast, accurate inspection
12 mm depth of field with fusion optics. Greenough optical design - provides very flat (plan) field of view and large depth of field
Complete apochromatic corrected microscopy system - 35° angle eye tubes - for comfort and convenience
122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
Resolution up to 250 lp/mm (without supplementary lens, 10× eyepieces), maximum resolution up to 500 lp/mm (2× supplementary lens, 10× eyepieces)
Fully encoded zoom - inspection at specific magnification can be easily repeated
Antimicrobial PC/ABS housing with AgTreat™
Large range of supplementary lenses as well as eyepieces
Adapt the optical performance to the application requirements
Integrated digital color camera with CMOS sensor
Possible to operate with 4k HDMI monitor using Enersight OSD
High definition live image 4k, 60fps, HDR
JPG images with up to 12 MP
Full HD MJPG video recording with 30 fps
Camera control via Windows using enersight desktop or via tablet/smartphone via enersight mobile application
Capture images directly to USB stick
USB-C connection to PC for enersight desktop
Delivery information: Includes Ivesta 3 (integrated camera); eyepiece 10×/23B, adj., eyeglasses x²; focus column for S-line; incident light base, small; sub base with reflector; KL300 LED fiberoptic light source w/ps; KL300 LED bracket for routine focusdrive; universal light guide, 600 mm; dust cover antistatic, small