Fluorescence Microscope for Cell Culture with Documentation, DMIL, Leica Microsystems
Supplier: Leica Microsystems
8122662
77676-394EA 30063.09 USD
77676-394
Fluorescence Microscope for Cell Culture with Documentation, DMIL, Leica Microsystems
Microscopes
The Leica DM IL LED enables phase contrast and fluorescence imaging for cell analysis. Its Leica K3C camera allows seamless documentation of routine tasks in cell culture labs, including confluency and transfection efficiency checks.
  • Easy-to-use operation that requires minimal training and maintenance
  • Cool, color-safe LED illumination for constant color temperature
  • Easy phase contrast: 10×, 20×, and 40× (not included) objectives share the same light ring
  • World class Leica objectives that provide brilliant images
  • Easily capture images of your sample with included camera
The Leica DM IL LED, equipped with a Leica K3C camera, is tailored for phase contrast and fluorescence imaging of cell cultures and individual cells. The system and the intuitive LASX software document seamlessly routine tasks in cell culture labs like confluence estimation and transfection efficiency checks. Elevate your cell culture research with the Leica DM IL LED, featuring a color camera for seamless image capture and providing fluorescence capabilities for enhanced visualisation and insights into dynamic cell culture phenomena.
Features and specifications include:
Leica DMIL LED inverted routine microscope with fixed stage
Microscope platform
Leica DM IL LED fluorescence
Incident light fluorescence
Transmitted Light
Slider for lightrings S40, complete
Objective: Obj. HI plan CY 10×/0.25 PH1
Documentation port
Trinocular tube HC ILT
Eyepiece HC plan 10×/20 BR
Eyepiece HC plan 10×/20 BR.M
Delivery information: Includes: Leica DM IL LED fluo; condenser S40/0.45; slider for lightrings S40, complete; Obj. HI plan CY 10×/0.25 PH1; trinocular tube HC ILT; eyepiece HC PLAN 10×/20 BR.; eyepiece HC PLAN 10×/20 BR.M; dust cover for DM IL, ILM; Leica K3C (color); C-mount HC 0.55x; pe 300 white MB; fluid light guide Ø3×2000 mm/series 380; lightguide coupler 1"; adapter sleeve for DM IL and DM1000; filter system GFP ET, s; obj. HI PLAN 4×/0.10 PH0; Obj.HI PLAN I 20×/0.30 PH1; Obj. HC PL FL L 40×/0.60 CORR PH2.