Educational Microscope with Koehler Illumination, DM750
Supplier: Leica Microsystems
8122661
77676-392EA 18512.53 USD
77676-392
Educational Microscope with Koehler Illumination, DM750
Microscopes
The Leica DM IL LED provides phase contrast and fluorescence monitoring for cell cultures, with LED illumination and precise optics. It supports routine tasks like confluence estimation and transfection efficiency checks, enhancing cell culture research with fluorescence capabilities.
  • Easy-to-use operation that requires minimal training and maintenance
  • Cool, color-safe LED illumination for constant color temperature
  • Easy phase contrast: 10×, 20×, and 40× (not included) objectives share the same light ring
  • World class Leica objectives that provide brilliant images
The Leica DM IL LED is tailored for phase contrast and fluorescence monitoring of cell cultures and individual cells. With its advanced LED illumination system and precision optics, it supports routine tasks in cell culture labs like confluence estimation and transfection efficiency checks. Elevate your cell culture research with the Leica DM IL LED, providing fluorescence capabilities for enhanced visualisation and insights into the dynamic of cell cultures. Inverted microscope for incident fluorescence with transmitted light LED illumination for bio-medicine applications.
Features and specifications include:
A fixed stage on three point support
Coaxial double knob for coarse and fine focus on both sides
A four-fold revolving nosepiece with 25×075 objective thread
Built-in fluorescence illumination axis with manual shutter
Fluorescence slider with three positions for filter blocks
Infinity optics with tube lens 1×
Interface for exchangeable observation tube and phototubes
Condenser holder stepless adjustable, including collector, aperture diaphragm, and filter holder
Illuminated on/off switch
IMC-interface
Power supply for TL LED illumination, including mains cable
Delivery information: Includes Leica DMIL LED inverted routine fluorescence; microscope with fixed stage; condenser S80/0.30; slider for light rings S80, complete; obj. HI plan CY 10×/0.25 PH1; binocular tube HC ILB; eyepiece HC plan 10×/20 BR; eyepiece HC plan 10×/20 BR.M; dust cover for DM IL, ILM; pe 300 white MB; fluid light guide Ø3×2000 mm/series 380; lightguide coupler 1"; adapter sleeve for DM IL and DM1000; filter system GFP ET; obj. HI plan 4×/0.10 PH0; obj.HI plan I 20×/0.30 PH1.