Magnesium chloride 25 mM

Supplier: EMPIRICAL BIOSCIENCE

MGCL2-25-25ML
77676-326EA 52.52 USD
77676-326
Magnesium chloride 25 mM
Magnesium chloride
Formula: MgCl₂
Storage Temperature: Freezer
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR