Phosphate Instrumental Test Kit
Supplier: CHEMetrics
K-8513
77674-860EA 42 USD
77674-860
Phosphate Instrumental Test Kit
Environmental Test Kits
  • Range: V-2000: 0 to 8.00 ppm / V-3000 and Spec: 0 to 5.00 ppm
  • Method: Stannous chloride
Delivery information: Phosphate test kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, activator solution, 25 ml sample cup, sample cup top, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.