CHEMets® Chlorine Visual HR Test 0 to 25 ppm
Supplier: CHEMetrics
K-2520D
77674-760EA 82 USD
77674-760
CHEMets® Chlorine Visual HR Test 0 to 25 ppm
Environmental Test Kits
Delivery information: The K-2520D chlorine HR CHEMets® test kit (0 to 25 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): R to 2500 CHEMets®refill, C to 2506 Comparato r, A to 2500 activato r solution, 25 ml sample cup, A to 0104 10ml syringe, and instructions.