7-(2-Hydroxyethyl)theophylline ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-(2-Hydroxyethyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Etofylline

A851165-1MG
77674-258EA 5 USD
77674-258
7-(2-Hydroxyethyl)theophylline ≥98%
7-(2-Hydroxyethyl)theophylline

Caution: for Research Use Only

Formula: C₉H₁₂N₄O₃
MW: 224.216 g/mol
MDL Number: MFCD00055055
CAS Number: 519-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR