4-Bromo-3-picoline hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methylpyridine hydrochloride

A340007-25G A340007-5G A340007-1G
77672-732EA 216 USD
77672-732 77672-734 77872-502
4-Bromo-3-picoline hydrochloride ≥95%
4-Bromo-3-picoline hydrochloride

Caution: for Research Use Only

Formula: C₆H₇BrClN
MDL Number: MFCD04966778
CAS Number: 40899-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR