2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine-2-carboxylic acid methylamide, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

A248099-1G
77672-218EA 49 USD
77672-218
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥98%
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
MW: 262.11 g/mol
MDL Number: MFCD08458483
CAS Number: 945863-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR