AZD-8055 98%

Supplier: Ambeed
Warning

Synonyms: (5-(2,4-Bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol, AZD 8055, (5-(2,4-bis((3s)-3-methylmorpholin-4-yl)pyrido(2,3-d)pyrimidin-7-yl)-2-methoxyphenyl)methanol, AZD8055, CCG168

A224432-10MG A224432-1MG A224432-100MG A224432-250MG A224432-1G A224432-5MG
76874-056EA 37 USD
76874-056 76874-058 77671-982 77671-984 77883-579 77883-580
AZD-8055 98%
AZD-8055

Caution: for Research Use Only

Formula: C₂₅H₃₁N₅O₄
MDL Number: MFCD16660191
CAS Number: 1009298-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR