D-(-)-Serine 95%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-Serine, H-D-Ser-OH

A111308-100G A111308-10G A111308-1G A111308-25G A111308-500G A111308-5G A111308-1000G
103823-628EA 6 USD
103823-628 76970-798 76970-800 76970-802 76970-804 76970-806 77284-104 77670-578
D-(-)-Serine 95%
D(-)-Serine

Caution: for Research Use Only

Formula: C₃H₇NO₃
Boiling Pt: 394.8 °C (760 mm Hg)
Melting Pt: 220 °C
Flash Pt: 192.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004269
CAS Number: 312-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR