Fluorescein-5-maleimide 95%
Supplier: Ambeed
|
Warning
|
A633839-25MG
A633839-250MG
A633839-100MG
A633839-10MG
76806-234EA
97.01
USD
76806-234
77569-208
77669-720
77669-722
Fluorescein-5-maleimide 95%
Fluorescein-5-maleimide
Caution: For research use only.
Formula:
C₂₄H₁₃NO₇ |
MDL Number:
MFCD00077345 CAS Number: 75350-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...