Fluorescein-5-maleimide 95%

Supplier: Ambeed
Warning

A633839-25MG A633839-250MG A633839-100MG A633839-10MG
76806-234EA 97.01 USD
76806-234 77569-208 77669-720 77669-722
Fluorescein-5-maleimide 95%
Fluorescein-5-maleimide
Formula: C₂₄H₁₃NO₇
MDL Number: MFCD00077345
CAS Number: 75350-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR