Tin(IV) oxide 99%

Supplier: Ambeed
Warning

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

A621734-500G A621734-25G
76790-670EA 74 USD
76790-670 77669-678
Tin(IV) oxide 99%
Tin (IV) oxide

Caution: for Research Use Only

Formula: O₂Sn
MDL Number: MFCD00011244
CAS Number: 18282-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR