n-Octacosane ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
N-octacosane
A553283-5G
A553283-100G
A553283-1G
A553283-25G
77602-342EA
28.7
USD
77602-342
77669-438
77669-440
77880-192
n-Octacosane ≥98%
n-Octacosane
Caution: for Research Use Only
Formula:
H₃C(CH₂)₂₆CH₃ MW: 394.77 g/mol Boiling Pt: 432 |
MDL Number:
MFCD00009355 CAS Number: 630-02-4 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...