2-(Methoxycarbonyl)-4-pyridineboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

A412065-5G A412065-1G A412065-100MG A412065-10G
77668-866EA 263 USD
77668-866 77872-654 77872-652 77872-653
2-(Methoxycarbonyl)-4-pyridineboronic acid pinacol ester ≥95%
2-(Methoxycarbonyl)-4-pyridineboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MDL Number: MFCD08752647
CAS Number: 957062-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR