Suramin hexasodium 98%
Supplier: Ambeed
|
Warning
|
A397022-1MG
A397022-100MG
A397022-50MG
A397022-1G
A397022-250MG
76788-912EA
42
USD
76788-912
77668-826
77668-828
77872-622
77872-623
Suramin hexasodium 98%
Suramin hexasodium
Caution: for Research Use Only
Formula:
C₅₁H₃₄N₆Na₆O₂₃S₆ |
MDL Number:
MFCD00210217 CAS Number: 129-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...