Neutral red 98% BS

Supplier: Ambeed
Warning

A375451-25G A375451-100G
76724-824EA 11 USD
76724-824 77668-742
Neutral red 98% BS
Neutral red

Caution: for Research Use Only

Formula: C₁₅H₁₇ClN₄
MDL Number: MFCD00012651
CAS Number: 553-24-2
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR