Saccharin sodium dihydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide dihydrate, Sodium Saccharin, Dihydrate, Saccharin sodium salt, o-sulphobenzimide sodium salt dihydrate, Saccharin Sodium Salt Dihydrate, o-Benzoic sulphimide sodium salt, o-sulfobenzimide sodium salt dihydrate, o-Benzoic sulfimide sodium salt

A361077-25G
77668-680EA 7 USD
77668-680
Saccharin sodium dihydrate ≥98%
Saccharin sodium dihydrate

Caution: for Research Use Only

Formula: C₇H₅NNaO₃S·2H₂O
MW: 242.21 g/mol
MDL Number: MFCD00151213
CAS Number: 6155-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR