Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride ≥97%

Supplier: Ambeed
Warning

A317143-1G A317143-250MG A317143-100MG A317143-50MG
77668-402EA 571 USD
77668-402 77668-404 77872-445 77872-446
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride ≥97%
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride

Caution: for Research Use Only

Formula: C₂₁H₂₇ClN₂O₃S
MDL Number: MFCD00058089
CAS Number: 69861-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR