(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: D-Phenylalanine tert-butyl ester hydrochloride, H-D-Phe-OtBu·HCl, H-D-Phe-OtBu.HCl

A246896-10G A246896-1G A246896-5G A246896-100G A246896-25G
76801-462EA 22 USD
76801-462 76801-464 76801-466 77600-992 77668-010
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 97%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride

Caution: for Research Use Only

Formula: C₁₃H₁₉NO₂·HCl
MDL Number: MFCD00153455
CAS Number: 3403-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR