Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 95%

Supplier: Ambeed
Warning

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

A223874-1G A223874-250MG A223874-25G A223874-5G A223874-100G
76786-070EA 8 USD
76786-070 76786-072 77600-796 77600-798 77667-856
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 95%
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

Caution: for Research Use Only

Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
MDL Number: MFCD00044310
CAS Number: 19817-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR