TAE684 ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-N4-(2-(isopropylsulphonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine, NVP-TAE 684, NVP-TAE684, 5-Chloro-N4-(2-(isopropylsulfonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine
A215396-100MG
A215396-10MG
A215396-250MG
A215396-50MG
77600-740EA
453.05
USD
77600-740
77600-742
77600-744
77667-804
TAE684 ≥98%
TAE684
Caution: for Research Use Only
Formula:
C₃₀H₄₀ClN₇O₃S |
MDL Number:
MFCD11977634 CAS Number: 761439-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...