Ethyl oleate ≥98%

Supplier: Ambeed
Warning

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

A193804-25G A193804-5G A193804-100G A193804-500G
77600-572EA 102.5 USD
77600-572 77600-574 77667-614 77667-616
Ethyl oleate ≥98%
Ethyl oleate

Caution: for Research Use Only

Formula: C₂₀H₃₈O₂
MDL Number: MFCD00009579
CAS Number: 111-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR