trans-Vaccenic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: trans-Octadec-11-enoic acid, Trans-11-Octadecenoic acid

A180388-100MG A180388-250MG A180388-1G
77667-490EA 210 USD
77667-490 77667-492 77871-902
trans-Vaccenic acid ≥98%
trans-Vaccenic acid

Caution: for Research Use Only

Formula: C₁₈H₃₄O₂
MW: 282.468 g/mol
MDL Number: MFCD00002734
CAS Number: 693-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR